Anti-P-EGFR Rabbit Polyclonal Antibody [clone: T669]
Catalog # BWRLBS4063
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:P-EGFR
- Clonality:Polyclonal
- Clone:T669
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human EGFR around the phosphorylation site of Threonine 669.
p-EGFR (T669) pAb detects endogenous levels of EGFR protein only when phosphorylated at Thr669 (removal of the signal peptide).
Type: Primary
Antigen: P-EGFR
Clonality: Polyclonal
Clone: T669
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat