Anti-hnRNP F Rabbit Polyclonal Antibody [clone: Y47]
Catalog # BWRLBS2014
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:hnRNP F
- Clonality:Polyclonal
- Clone:Y47
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 20-60 of Human hnRNP F.
hnRNP F (Y47) pAb detects endogenous levels of hnRNP F protein.
Type: Primary
Antigen: hnRNP F
Clonality: Polyclonal
Clone: Y47
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat