Anti-Neurophysin I Rabbit Polyclonal Antibody [clone: K40]
Catalog # BWRLBS3475
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Neurophysin I
- Clonality:Polyclonal
- Clone:K40
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 11-60 of Human Neurophysin I.
Neurophysin I (K40) pAb detects endogenous levels of Neurophysin I protein.
Type: Primary
Antigen: Neurophysin I
Clonality: Polyclonal
Clone: K40
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat