Anti-Ob-R Rabbit Polyclonal Antibody [clone: L662]
Catalog # BWRLBS3460
Leverantör: Bioworld Technology
Specifikationer
- Antikroppstyp:Primär
- Antigensymbol:Ob-R
- Klonalitet:Polyclonal
- Klon:L662
- Värd:Rabbit
- ImmunKemi:Yes
- Reaktivitet:Human
- Western blot:Yes
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Synthetic peptide, corresponding to amino acids 650-700 of Human Ob-R.
Ob-R (L662) pAb detects endogenous levels of Ob-R protein.
Type: Primary
Antigen: Ob-R
Clonality: Polyclonal
Clone: L662
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human