Anti-mGluR-2/3 Rabbit Polyclonal Antibody [clone: N867pAb ]
BWRLBS3430
- Antibody type:Primary
- Antigen symbol:mGluR-2/3
- Clonality:Polyclonal
- Clone:N867pAb
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 836-886 of Human mGluR-2.
mGluR-2/3 (N867) pAb detects endogenous levels of mGluR-2/3 protein
Type: Primary
Antigen: mGluR-2/3
Clonality: Polyclonal
Clone: N867pAb
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat