Anti-PFKP Rabbit Polyclonal Antibody [clone: E369]
Catalog # BWRLBS2911
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PFKP
- Clonality:Polyclonal
- Clone:E369
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 350-400 of Human PFKP.
PFKP (E369) pAb detects endogenous levels of PFKP protein.
Type: Primary
Antigen: PFKP
Clonality: Polyclonal
Clone: E369
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat