Anti-Apaf-1-ALT Rabbit Polyclonal Antibody [clone: H324]
Catalog # BWRLBS1016
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Apaf-1-ALT
- Clonality:Polyclonal
- Clone:H324
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 301-350 of Human Apaf-1-ALT.
APAF-1-ALT(H324) pAb detects endogenous levels of APAF-1 Isoform 6 (Apaf-1-ALT) protein.
Type: Primary
Antigen: Apaf-1-ALT
Clonality: Polyclonal
Clone: H324
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat