Order Entry
Sweden
ContactUsLinkComponent
Hood, open face, Tyvek®
Hood, open face, Tyvek®
Catalog # SSSAHB-TY01
Leverantör:  3S ARBEITSSCHUTZ
undefined
Hood, open face, Tyvek®
Catalog # SSSAHB-TY01
Leverantör:  3S ARBEITSSCHUTZ

Specifikationer

  • Garment application:
    Laboratory
  • Colour:
    White
  • Model:
    PH30L0
  • Pk:
    200

Specifikationer

Om denna produkt

Single use hood with shoulder cover. Made from breathable Tyvek®.