Order Entry
Sweden
Orders LinkContactUsLinkComponent
Microvolume syringes 10 µl
Microvolume syringes 10 µl
Certificates
Microvolume syringes 10 µl
Microvolume syringes 10 µl
18 Options Available Below
Product Details & Documents
These 10 µl metal plunger syringes with stainless steel needle are ideal for making microvolume injections of 1,5 to 8 µl. Each plunger is individually fitted for each microsyringe for perfect sealing.
  • Accuracy and reproducibility: ±1%
  • Borosilicate glass barrel OD: 6,5 mm and 8 mm for guided plunger
  • Scale length: 54,1 mm
Plunger protection incorporates a plunger protection guide and plunger button reinforcement to minimise the risk of bent plungers.
SuperfleX microsyringes use a super elastic alloy plunger that will not bend or kink.
Guided plunger microsyringes have an extended barrel and reinforced plunger that provides additional protection.
Product Family Options
Fixed needle, barrel OD 6,5 mm
Product Information
  • Gauge
    Volume
    Length
    Ext.Ø
    Type of tip
    Type
    Availability
    Price
  • 549-0429
    Plunger protection syringe
    Plunger protection syringe
    23
    10 µl
    42 mm
    0,63 mm
    Conical
    10F-4.2/0.63C
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1 400,00 kr
    1 400,00 kr
    1 * 1 Items
       1 400,00 kr
  • 549-0279
    Plunger protection syringe
    Plunger protection syringe
    26
    10 µl
    50 mm
    0,47 mm
    Bevelled
    10F
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 535,00 kr
    535,00 kr
    1 * 1 Items
       535,00 kr
  • 549-0280
    Plunger protection syringe
    Plunger protection syringe
    26
    10 µl
    50 mm
    0,47 mm
    Bevelled
    10F
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 2 675,00 kr
    2 675,00 kr
    1 * 6 Items
       2 675,00 kr
  • 548-0198
    Plunger protection syringe
    Plunger protection syringe
    26
    10 µl
    50 mm
    0,47 mm
    Bevelled
    10F
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 4 250,00 kr
    4 250,00 kr
    1 * 10 Items
       4 250,00 kr
  • 548-0199
    Plunger protection syringe
    Plunger protection syringe
    26
    10 µl
    50 mm
    0,47 mm
    Bevelled
    10F
    Log in for availability
    Log in for availability
    1 * 25 Items
    1 * 25 Items - 9 950,00 kr
    9 950,00 kr
    1 * 25 Items
       9 950,00 kr
  • 549-0431
    Plunger protection syringe
    Plunger protection syringe
    26
    10 µl
    50 mm
    0,47 mm
    Conical
    10F-5C
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 970,00 kr
    970,00 kr
    1 * 1 Items
       970,00 kr
  • 549-0284
    SuperfleX flexible plunger syringe
    SuperfleX flexible plunger syringe
    26
    10 µl
    50 mm
    0,47 mm
    Bevelled
    10FX
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 855,00 kr
    855,00 kr
    1 * 1 Items
       855,00 kr
  • 549-0285
    SuperfleX flexible plunger syringe
    SuperfleX flexible plunger syringe
    26
    10 µl
    50 mm
    0,47 mm
    Bevelled
    10FX
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 4 150,00 kr
    4 150,00 kr
    1 * 6 Items
       4 150,00 kr
  • 549-0434
    SuperfleX flexible plunger syringe
    SuperfleX flexible plunger syringe
    26
    10 µl
    50 mm
    0,47 mm
    Conical
    10FX-5C
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1 025,00 kr
    1 025,00 kr
    1 * 1 Items
       1 025,00 kr
  • 548-0934
    SuperfleX flexible plunger syringe
    SuperfleX flexible plunger syringe
    23
    10 µl
    50 mm
    0,63 mm
    Conical
    10FX-5/0.63C
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 4 900,00 kr
    4 900,00 kr
    1 * 6 Items
       4 900,00 kr
  • 548-0779
    SuperfleX flexible plunger syringe
    SuperfleX flexible plunger syringe
    26
    10 µl
    50 mm
    0,47 mm
    Conical
    10FX-5C
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 4 900,00 kr
    4 900,00 kr
    1 * 6 Items
       4 900,00 kr
  • 549-0430
    Plunger protection syringe
    Plunger protection syringe
    26
    10 µl
    70 mm
    0,47 mm
    Bevelled
    10F-7
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 735,00 kr
    735,00 kr
    1 * 1 Items
       735,00 kr
Fixed needle with guided plunger, barrel OD 8 mm
Product Information
  • Gauge
    Volume
    Length
    Ext.Ø
    Type of tip
    Type
    Availability
    Price
  • 549-0290
    Guided plunger syringe
    Guided plunger syringe
    26
    10 µl
    50 mm
    0,47 mm
    Bevelled
    10F-GP
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1 450,00 kr
    1 450,00 kr
    1 * 1 Items
       1 450,00 kr
Removable needle, barrel OD 6,5 mm
Product Information
  • Gauge
    Volume
    Length
    Ext.Ø
    Type of tip
    Type
    Availability
    Price
  • 549-0281
    Plunger protection syringe
    Plunger protection syringe
    26
    10 µl
    50 mm
    0,47 mm
    Bevelled
    10R
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 920,00 kr
    920,00 kr
    1 * 1 Items
       920,00 kr
  • 549-0283
    Plunger protection syringe
    Plunger protection syringe
    26
    10 µl
    50 mm
    0,47 mm
    Bevelled
    10R
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 4 850,00 kr
    4 850,00 kr
    1 * 6 Items
       4 850,00 kr
  • 549-0287
    SuperfleX flexible plunger syringe
    SuperfleX flexible plunger syringe
    26
    10 µl
    50 mm
    0,47 mm
    Bevelled
    10RX
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 5 975,00 kr
    5 975,00 kr
    1 * 6 Items
       5 975,00 kr
Removable needle with guided plunger, barrel OD 8 mm
Product Information
  • Gauge
    Volume
    Length
    Ext.Ø
    Type of tip
    Type
    Availability
    Price
  • 549-0291
    Guided plunger syringe
    Guided plunger syringe
    26
    10 µl
    50 mm
    0,47 mm
    Bevelled
    10R-GP
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1 725,00 kr
    1 725,00 kr
    1 * 1 Items
       1 725,00 kr
  • SGEA002450CAL
    Guided plunger syringe
    Guided plunger syringe
    26
    10 µl
    50 mm
    0,47 mm
    Bevelled
    10R-GP (Calibrated)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 4 325,00 kr
    4 325,00 kr
    1 * 1 Items
       4 325,00 kr
Replacement needles for 10 µl removable needle syringes
Product Information
  • Catalog #
    Type of tip
    Length
    Gauge
    Type
    Ext.Ø
    Availability
    Price
  • Conical
    50 mm
    23
    N10-5/0.63C
    0,63 mm
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 675,00 kr
    675,00 kr
    1 * 2 Items
       675,00 kr
Product Information
  • Catalog #
    Type of tip
    Length
    Gauge
    Type
    Ext.Ø
    Availability
    Price
  • Conical
    50 mm
    26
    N10-5C
    0,47 mm
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 675,00 kr
    675,00 kr
    1 * 2 Items
       675,00 kr
Product Information
  • Catalog #
    Type of tip
    Length
    Gauge
    Type
    Ext.Ø
    Availability
    Price
  • Conical
    70 mm
    26
    N10-7C
    0,47 mm
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 675,00 kr
    675,00 kr
    1 * 2 Items
       675,00 kr
Product Information
  • Catalog #
    Type of tip
    Length
    Gauge
    Type
    Ext.Ø
    Availability
    Price
  • Conical
    70 mm
    23
    N10-7/0.63C
    0,63 mm
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 675,00 kr
    675,00 kr
    1 * 2 Items
       675,00 kr
Product Information
  • Catalog #
    Type of tip
    Length
    Gauge
    Type
    Ext.Ø
    Availability
    Price
  • Bevelled
    115 mm
    26
    N10-11.5
    0,47 mm
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 1 100,00 kr
    1 100,00 kr
    1 * 5 Items
       1 100,00 kr
Product Information
  • Catalog #
    Type of tip
    Length
    Gauge
    Type
    Ext.Ø
    Availability
    Price
  • Bevelled
    50 mm
    23
    N10-7/0.63
    0,63 mm
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 730,00 kr
    730,00 kr
    1 * 5 Items
       730,00 kr
Product Information
  • Catalog #
    Type of tip
    Length
    Gauge
    Type
    Ext.Ø
    Availability
    Price
  • Bevelled
    70 mm
    26
    N10-7
    0,47 mm
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 1 100,00 kr
    1 100,00 kr
    1 * 5 Items
       1 100,00 kr
Product Information
  • Catalog #
    Type of tip
    Length
    Gauge
    Type
    Ext.Ø
    Availability
    Price
  • Side hole/dome
    50 mm
    26
    N10-5H
    0,47 mm
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 990,00 kr
    990,00 kr
    1 * 2 Items
       990,00 kr
Product Information
  • Catalog #
    Type of tip
    Length
    Gauge
    Type
    Ext.Ø
    Availability
    Price
  • On column
    100 mm
    -
    N10-OC-10/0.17VS
    0,17 mm
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 1 200,00 kr
    1 200,00 kr
    1 * 2 Items
       1 200,00 kr
Product Information
  • Catalog #
    Type of tip
    Length
    Gauge
    Type
    Ext.Ø
    Availability
    Price
  • On column
    75 mm
    -
    N10-OC-7.5/0.23
    0,23 mm
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 1 200,00 kr
    1 200,00 kr
    1 * 2 Items
       1 200,00 kr
Product Information
  • Catalog #
    Type of tip
    Length
    Gauge
    Type
    Ext.Ø
    Availability
    Price
  • LC
    51 mm
    22
    N10-LC
    0,028" mm
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 730,00 kr
    730,00 kr
    1 * 5 Items
       730,00 kr
Product Information
  • Catalog #
    Type of tip
    Length
    Gauge
    Type
    Ext.Ø
    Availability
    Price
  • Bevelled
    50 mm
    26
    N10-5
    0,47 mm
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 730,00 kr
    730,00 kr
    1 * 5 Items
       730,00 kr