Z-Val-Ala-Asp-(Ome)-fluoromethylketone, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Storage temperature: +4 °C
Specifications
- Storage conditions:+4 °C
- Protein/peptide name:Z-Val-Ala-Asp-(Ome)-Fluoromethylketone
- Molecular weight:467.487
- Molecular formula:C₂₂H₃₀FN₃O₇