Order Entry
Sweden
ContactUsLinkComponent
DCPIP assay reagent
DCPIP assay reagent
Catalog # CAYM700942-700
Supplier:  Cayman Chemical
CAS Number:  
DCPIP assay reagent
Catalog # CAYM700942-700
Supplier:  Cayman Chemical
CAS Number:  
Specifications
  • Pk:
    700 µG
  • Description:
    DCPIP assay reagent
Specifications
About this item
The vial contains 700 μg of 2,6-dichlorophenolindophenol (DCPIP) assay reagent dissolved in UltraPure water. Thaw the reagent prior to use in the MitoCheck® Complex II Activity Assay Kit.