Chemical resistant overalls, MICROGARD® 2500 PLUS, models 111 / 122
: Ansell
These overalls are made from a durable microporous PP laminate with a non woven inner layer. The fabric provides an excellent barrier to chemical spray and infective agents.
- 3-piece hood is designed for optimum fit with respirators, particularly full face masks
- Elasticated hood, wrist, waist and ankles
- 2-way front zip with resealable storm flap
- Finger loops to prevent sleeve movement when working above the head
- Stitched and taped seams: Increased strength and an effective liquid & particle barrier
- Moisture vapour permeable ('breathable') to help reduce the risk of heat stress
- Low linting - Reduced risk of contamination in critical areas
Typical applications include: Viral contaminated areas (including Avian Influenza), biological protection, emergency medical response, medical research, chemical and pharmaceutical industries, low pressure industrial cleaning, industrial paint spraying, nuclear industry.
: Type 3-B/4-B/5-B/6-B, EN 14126, EN 1073-2, EN 1149-5; Cat. III
- Catalog No:
- ANSEWH25T-00122-06
- ANSEWH25T-00122-05
- ANSEWH25T-00122-04
- ANSEWH25T-00111-07
- ANSEWH25T-00122-03
- ANSEWH25T-00122-02
- ANSEWH25T-00111-06
- ANSEWH25T-00111-05
- ANSEWH25T-00111-04
- ANSEWH25T-00111-03
- ANSEWH25T-00122-07
- Model:
- 122
- 122
- 122
- 111
- 122
- 122
- 111
- 111
- 111
- 111
- 122
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial
- Chemical protection, Medical, Laboratory, Clinical, Industrial