Order Entry
Sweden
ContactUsLinkComponent
Sodium hydroxide 95.0-100.5%, pellets FCC, NF, J.T.Baker®
Corrosive
Sodium hydroxide 95.0-100.5%, pellets FCC, NF, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Sodium hydroxide 95.0-100.5%, pellets FCC, NF, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Food GMP Manufactured Product:
    Passes Test
  • GMP Manufactured Product:
    ≥11.0
  • Meets N.F. Requirements:
    Passes Test
  • Meets F.C.C. Requirements:
    ≤0.5%
  • CAUTION: For Manufacturing, processing or repackaging:
    95.0 - 100.5%
  • Bulk Food Chemical:
    54.0 - 59.8%
  • Bulk Pharmaceutical Chemical:
    ≤3.0%
  • NF - Identification A (Sodium):
    Passes Test
  • NF - Identification B (pH):
    ≥11.0
  • NF - Insoluble Substances and Organic Matter:
    Passes Test
  • NF - Potassium (K):
    ≤0.5 %
  • NF - Total Alkali:
    95.0 - 100.5 %
  • NF - Content of Sodium:
    54.0 - 59.8 %
  • NF - Carbonate (as Na₂CO₃):
    ≤3.0 %
  • FCC - Insoluble Substances and Organic Matter:
    Passes Test
  • FCC - Assay:
    95.0 - 100.5 %
  • FCC - Carbonate (as Na₂CO₃):
    ≤3.0 %
  • FCC - Arsenic (As):
    ≤3 ppm
  • FCC - Lead (Pb):
    ≤2 mg/kg
  • FCC - Mercury (Hg):
    ≤0.1 ppm

Specification Test Results

Related Information