Water Ph. Eur., BP, pharma grade
Catalog # APPC141074.1315
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pk:10 L
- Seq:5214
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
About this item
:
:18.02 g/mol
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332