Syringes for CTC analytics systems
Supplier: Trajan Scientific and Medical
Jump to:
About this item
* Gas-tight.
46 Options Available Below
Product Details & Documents
* Gas-tight.
Product Family Options
For CTC analytics HTS, HTC and LC PAL instruments
Product Information
- GaugeVolumeLengthExt.ØType of tipTypeAvailabilityPrice
- 554-1444Fixed needle
- 554-1452Fixed needle
- 548-1625Fixed needle
- 548-1630Removable needleRemovable needle22100 µl51 mm0,72 mmLC100R-C/T-GT-LC *Log in for availability1 * 1 Items1 475,00 kr
- 548-1631Fixed needle
- 554-1457Fixed needle
- 554-1458Fixed needleFixed needle22100 µl51 mm0,72 mmLC100F-CTC-GT-LC (0.4) *Log in for availability1 * 1 Items1 500,00 kr
- 548-1643Removable needleRemovable needle22250 µl51 mm0,72 mmLC250R-C/T-GT-LC *Log in for availability1 * 1 Items1 425,00 kr
- 554-1464Fixed needleFixed needle22500 µl51 mm0,72 mmLC500F-CTC-GT-LC (0.4) *Log in for availability1 * 1 Items1 525,00 kr
- 554-1465Removable needleRemovable needle221000 µl51 mm0,72 mmLC1MR-CTC-GT-LC(0.4) *Log in for availability1 * 1 Items1 500,00 kr
For CTC analytics headspace, CombiPAL and GC PAL
Product Information
- GaugeVolumeLengthExt.ØType of tipTypeAvailabilityPrice
- 548-1664Fixed needleFixed needle261000 µl56 mm0,47 mmSide hole1MF-CTC-EGT-HS-5.6/0.47H *Log in for availability1 * 1 Items2 575,00 kr
- 548-1663Fixed needleFixed needle231000 µl56 mm0,63 mmSide hole1MF-CTC-EGT-HS-5.6/0.63H *Log in for availability1 * 1 Items2 575,00 kr
- 548-1669Fixed needleFixed needle262500 µl56 mm0,47 mmSide hole2.5MF-CTC-EGT-HS-5.6/0.47H *Log in for availability1 * 1 Items2 475,00 kr
- 548-1668Fixed needleFixed needle232500 µl56 mm0,63 mmSide hole2.5MF-CTC-EGT-HS-5.6/0.63H *Log in for availability1 * 1 Items2 475,00 kr
For CTC analytics CombiPAL and GC PAL instruments
Product Information
- GaugeVolumeLengthExt.ØType of tipTypeAvailabilityPrice
- 558-0611Removable needleRemovable needle260,5 µl50 mm0,47 mmConical0.5BNR-C/T-5/0.47CLog in for availability1 * 1 Items2 125,00 kr
- 558-0612Removable needleRemovable needle230,5 µl50 mm0,63 mmConical0.5BNR-C/T-5/0.63CLog in for availability1 * 1 Items2 125,00 kr
- SGEA000790Removable needleRemovable needle232 µl50 mm0,63 mmConical2BR-C/T-5/0.63CLog in for availability1 * 1 Items2 000,00 kr
- SGEA0019821Fixed needle
- 548-1583Fixed needle
- SGEA0019811Fixed needle
- 549-0403Fixed needle
- SGEA001984Removable needleRemovable needle235 µl50 mm0,63 mmConical5R-C/T-0.63CLog in for availability1 * 1 Items1 575,00 kr
- 554-1442Fixed needleFixed needle2610 µl50 mm0,47 mmConical10F-CTC-5/0.47CLog in for availability1 * 1 Items1 100,00 kr
- 554-1449Fixed needleFixed needle2610 µl50 mm0,47 mmConical10F-C/T-GT-5/0.47C 6 pack *Log in for availability1 * 6 Items5 650,00 kr
- SGEA0029771Fixed needleFixed needle2610 µl50 mm0,47 mmConical10F-BT-GT-5/0.47CLog in for availability1 * 1 Items1 025,00 kr
- 554-1450Fixed needleFixed needle2610 µl50 mm0,47 mmConical10F-C/T-GT-5/0.47C *Log in for availability1 * 1 Items1 025,00 kr
- SGEA0029781Fixed needleFixed needle2610 µl70 mm0,47 mmConical10F-BT-7/0.47C SYRINGELog in for availability1 * 1 Items1 125,00 kr
- 548-1561Removable needleRemovable needle2610 µl50 mm0,47 mmConical10R-BT-GT-0.47C *Log in for availability1 * 1 Items1 675,00 kr
- 548-1608Removable needleRemovable needle2610 µl50 mm0,47 mmConical10R-C/T-GT-5/0.47C *Log in for availability1 * 1 Items1 675,00 kr
- 548-1938Fixed needleFixed needle2610 µl50 mm0,47 mmConical10F-BT-5/0.47C 6 packLog in for availability1 * 6 Items4 775,00 kr
- 548-1937Fixed needleFixed needle2610 µl50 mm0,47 mmConical10F-C/T-5/0.47C 6 packLog in for availability1 * 6 Items4 775,00 kr
- SGEA0029671Fixed needle
- 558-0610Fixed needle
- SGEA002971Fixed needleFixed needle2310 µl50 mm0,63 mmConical10F-C/T-5/0.63C 6 packLog in for availability1 * 6 Items5 200,00 kr
- 554-1448Fixed needleFixed needle2310 µl50 mm0,63 mmConical10F-C/T-GT-5/0.63C 6 packLog in for availability1 * 6 Items6 775,00 kr
- 562-1043Fixed needleFixed needle2310 µl50 mm0,63 mmConical10F-C/T-GT-5/0.63C *Log in for availability1 * 1 Items1 400,00 kr
- SGEA0029741Fixed needle
- 548-1607Fixed needleFixed needle2210 µl50 mm0,72 mmSide hole10F-CTC-GT-5/0.72H *Log in for availability1 * 1 Items1 175,00 kr
- 554-1451Fixed needleFixed needle2625 µl50 mm0,47 mmConical25F-CTC-GT-5/0.47C *Log in for availability1 * 1 Items1 775,00 kr
- 548-1620Removable needleRemovable needle2625 µl50 mm0,47 mmSide hole25R-C/T-GT-5/0.47H *Log in for availability1 * 1 Items2 075,00 kr
- 548-1939Fixed needleFixed needle2325 µl50 mm0,63 mmConical25F-BT-GT-0.63CLog in for availability1 * 1 Items1 525,00 kr
- 548-1619Fixed needleFixed needle2325 µl50 mm0,63 mmConical25F-C/T-GT-5/0.63C *Log in for availability1 * 1 Items1 525,00 kr
- 548-1632Removable needleRemovable needle26100 µl50 mm0,47 mmConical100R-C/T-GT-5/0.47C *Log in for availability1 * 1 Items1 800,00 kr
- 554-1456Fixed needleFixed needle26100 µl50 mm0,47 mmConical100F-CTC-GT-5/0.47C *Log in for availability1 * 1 Items1 775,00 kr
- 548-1633Fixed needleFixed needle23100 µl50 mm0,63 mmConical100F-C/T-GT-5/0.63C *Log in for availability1 * 1 Items1 725,00 kr
- 554-1461Fixed needleFixed needle26250 µl50 mm0,47 mmConical250F-CTC-GT-5/0.47C *Log in for availability1 * 1 Items1 650,00 kr
Replacement needles
Product Information
- Catalog #Type of tipExt.ØLengthForGaugeDescriptionAvailabilityPrice
Replacement plunger
Product Information
- Catalog #ForDescriptionAvailabilityPrice