Accessories for SCAT Safety Caps
Supplier: S.C.A.T.
Certificates
About this item
Helpful supplies for the safe use of SCAT and SymLine products Safety Caps and Safety Waste Caps.
- SCAT connectors are available in the following versions:
- Conical connectors
- Y-connectors
- Tube connectors
- Blind plugs
- Capillary connector
SCAT thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters, or other disposal and supply units.
Related Information
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
TDS Universal Waste Hub Jan