0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Amiloride hydrochloride
Amiloride hydrochloride ≥98%
Amiloride hydrochloride ≥98%
:
Cayman Chemical
Amiloride hydrochloride ≥98%
:
Cayman Chemical
:
2016-88-8
:
17440-83-4
:
C₆H₈ClN₇O·HCl
:
MFCD00058197
:
266.09 g/mol