Potato native acid phosphatase
Katalog # 524529-1
Leverantör: Merck Millipore (Calbiochem)
CAS-nummer:
Specifikationer
- Pk:1 KU
- CAS No.:9001-77-8
- Enzyme type:Native
- Species:Potato
- Storage conditions:−2...8 °C
- Form:Lyophilized powder
- Enzyme name:Acid phosphatase
- Enzyme activity:≥2 units/mg protein
- Solubility:3,2 M ammonium sulphate, pH 6,0, containing 1% BSA or 6% BSA in H₂O
Specifikationer
Om denna produkt
Native potato acid phosphatase. Ideal for the removal of phosphate groups from phosphoproteins, such as β-casein, ovalbumin and pepsinogen.
One unit is defined as the amount of enzyme that will hydrolyse 1 µmol of pNPP/min at 25 °C, pH 5,6.