Order Entry
Sweden
ContactUsLinkComponent
Organic vapour monitors
Organic vapour monitors
Catalog # 514-0007
Supplier:  3M
undefined
Organic vapour monitors
Catalog # 514-0007
Supplier:  3M

Specifications

  • Description:
    Ethylene oxide monitors
  • Seq:
    01
  • Pk:
    5

Specifications

About this item

Monitor uses the proven scientific principle of diffusion which involves the gradual mixing of molecules of two or more substances for measuring personal exposure in work areas of industries.

  • Ethylene oxide monitor utilises brominated carbon to collect and convert the reactive ethylene oxide to a more stable chemical derivative, 2-bromoethanol
  • Compound then desorbed with methylene chloride in methanol
  • Accuracy within ±25% for 8-hour samplings at 1 ppm
  • Accuracy within ±35% for 8-hour samplings at the action limit

18-month shelf life.

Certifications: OSHA