Order Entry
Sweden
ContactUsLinkComponent
Anti-F4/80 Rat Monoclonal Antibody (APC-Cyanine7) [clone: BM8.1]
Anti-F4/80 Rat Monoclonal Antibody (APC-Cyanine7) [clone: BM8.1]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-F4/80 Rat Monoclonal Antibody (APC-Cyanine7) [clone: BM8.1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 25-4801-U100
  • 25-4801-U025
  • Antigen symbol:
  • EMR1, Ly71
  • EMR1, Ly71
  • Antigen name:
  • F4/80
  • F4/80
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)/Cy7®
  • APC (Allophycocyanin)/Cy7®
  • Clone:
  • BM8.1
  • BM8.1
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 13733
  • 13733
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • 2-8°C protected from light

Specifications

Related Information