Order Entry
Sweden
ContactUsLinkComponent
Anti-NS1 Mouse Monoclonal Antibody [clone: M86642]
Anti-NS1 Mouse Monoclonal Antibody [clone: M86642]
Catalog # ABNOMAB14220
Supplier:  Abnova
Anti-NS1 Mouse Monoclonal Antibody [clone: M86642]
Catalog # ABNOMAB14220
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    NS1
  • Clonality:
    Monoclonal
  • Clone:
    M86642
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Virus
  • Form:
    Liquid
  • Storage buffer:
    In 0.015 M potassium phosphate, 0.85% NaCl, pH 7.2 (0.05% sodium azide).
  • Immunogen:
    Recombinant protein corresponding to amino acids 792-1144 of Zika virus NS1.
  • Purification:
    DEAE column chromatography
  • Size:
    500 µG
  • Pk:
    500 µG

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant Zika virus NS1.

Type: Primary
Antigen: NS1
Clonality: Monoclonal
Clone: M86642
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Viruses