Silicone antifoam, Supelco®
Catalog # 1.07743.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Identity (IR-spectrum):Conforms
- Density (d 20/4):0,98 - 1,01 g/ml
- Application Test:Conforms
Specification Test Results
About this item
:
Boiling Pt:300 °C (1013 hPa)
Density:0.98…1.01 g/ml
:M-Clarity™ quality level = MQ100