Order Entry
Sweden
ContactUsLinkComponent
Organelle-ID™ RGB reagent IV
Organelle-ID™ RGB reagent IV
Catalog # ENZOENZ53009C200
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Organelle-ID™ RGB reagent IV
Catalog # ENZOENZ53009C200
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    200 µl
  • Description:
    Reagent IV, Organelle-ID RGB® for 1000 assays
  • Seq:
    02
  • Purity:
    ≥93 % (HPLC)
  • Concentration:
    500X

Specifications

About this item

Organelle-ID RGB® reagent IV is a mixture of cell-permeable red fluorescent endoplasmic reticulum dye, green fluorescent lysosomal dye, and blue fluorescent nucleic acid dye.

To visualise of the target organelles by fluorescence/confocal microscopy.