Order Entry
Sweden
ContactUsLinkComponent
Human Syntaxin 1A (from E. coli)
Human Syntaxin 1A (from E. coli)
Supplier:  Biorbyt
Certificates
Human Syntaxin 1A (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    STX1 although stable 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please prevent freeze-thaw cycles.
  • Protein synonyms:
    HPC 1
  • Protein/peptide name:
    Syntaxin 1A
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Sequence:
    MKDRTQELRT AKDSDDDDDV AVTVDRDRFM DEFFEQVEEI RGFIDKIAEN VEEVKRKHSA ILASPNPDEK TKEELEELMS DIKKTANKVR SKLKSIEQSI EQEEGLNRSS ADLRIRKTQH STLSRKFVEV MSEYNATQSD YRERCKGRIQ RQLEITGRTT TSEELEDMLE SGNPAIFASG IIMDSSISKQ ALSEIETRHS EIIKLENSIR ELHDMFMDMA MLVESQ.
  • Formulation:
    The protein solution contains 20 mM Tris-HCl pH7.5, 10% glycerol, and 1mM DTT.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together