Rat recombinant IL2
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Species:Rat
- Storage conditions:Store at −20 °C.
- Biological activity:The ED50 as determined by the dose-dependent stimulation of the proliferation of Mouse CTLL-2 cells is < 0.4 ng/ml, corresponding to a specific activity of > 2.5 x 106 units/mg.
- Protein synonyms:TCGF|T-cell growth factor
- UniProtKB:P17108
- Protein/peptide name:IL2
- Purity:>98%
- Solubility:It is recommended to reconstitute the lyophilized Interleukin in sterile 100mM AcOH not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAIAQSI ISTM
- Formulation:The protein (1mg/ml) was lyophilized without any additives.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together