Human recombinant ADK (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Adenosine Kinase Human although stable at 4 °C for weeks, should be stored desiccated below −18 °C. Please prevent freeze thaw cycles.
- Protein synonyms:Adenosine 5''-phosphotransferase
- Protein/peptide name:ADK
- Purity:Greater than 95% as determined by SDS-PAGE.
- Sequence:MGSSHHHHHH SSGLVPRGSH MRENILFGMG NPLLDISAVV DKDFLDKYSL KPNDQILAED KHKELFDELV KKFKVEYHAG GSTQNSIKVA QWMIQQPHKA ATFFGCIGID KFGEILKRKA AEAHVDAHYY EQNEQPTGTC AACITGDNRS LIANLAAANC YKKEKHLDLE KNWMLVEKAR VCYIAGFFLT VSPESVLKVA HHASENNRIF TLNLSAPFIS QFYKESLMKV MPYVDILFGN ETEAATFARE QGFETKDIKE IAKKTQALPK MNSKRQRIVI FTQGRDDTIM ATESEVTAFA VLDQDQKEII DTNGAGDAFV GGFLSQLVSD KPLTECIRAG HYAASIIIRR TGCTFPEKPD FH.
- Formulation:ADK Human solution containing 20 mM Trsi HCL pH-8, 1mM DTT, 1mM EDTA, 50mM NaCl 20% glycerol.
- Tested applications:SDS-PAGE
Frequently Bought Together