Order Entry
Sweden
ContactUsLinkComponent
Human recombinant CDK4 (from E. coli)
Human recombinant CDK4 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant CDK4 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
  • Protein synonyms:
    Cell division
  • Protein/peptide name:
    CDK4
  • Purity:
    Greater than 95% as determined by SDS-PAGE. Single band on Western Blot.
  • Formulation:
    CDK4 is supplied in 10mM Bis-Tris Propane pH9.0 and 50% Glycerol.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together