Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteiner
D-Dimer
D-Dimer
D-Dimer
Catalog # BIRBORB79661-1
Supplier:
Biorbyt
CAS Number:
Expand image
D-Dimer
Catalog # BIRBORB79661-1
Supplier:
Biorbyt
CAS Number:
Specifications
Förp.:
1 mg
Storage conditions:
Store at -2 to -8 °C.
Protein/peptide name:
D-Dimer
Renhet:
Greater than 95% pure SDS-PAGE. Protein G Chromatography
Specifications
About this item
Produktinformation
Dokumentation
Produktinformation
Dokumentation