Order Entry
Sweden
ContactUsLinkComponent
Human TNFSF1A (from E. coli)
Human TNFSF1A (from E. coli)
Supplier:  Biorbyt
Certificates
Human TNFSF1A (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at less than 20 °C. though stable at room temperature for 3 weeks.
    Reconstituted protein solution can be stored at 4-7 °C. for 2-7 days.
    Aliquots of reconstituted samples are stable at less than 20 °C. for 3 months.
  • Protein/peptide name:
    TNFSF1A
  • Purity:
    Greater than 95% as determined by SEC-HPLC and reducing SDS-PAGE.
  • Sequence:
    First Nter minal amino acids is Met
  • Formulation:
    Lyophilized from a 0.2 uM filtered solution of 20mM PB, 300mM NaCl, pH7.4
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together