Order Entry
Sweden
ContactUsLinkComponent
Human recombinant PTP1C (from E. coli)
Human recombinant PTP1C (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant PTP1C (from E. coli)
Supplier:  Biorbyt

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    TyrosineProtein Phosphatase NonReceptor Type 6
  • UniProtKB:
    P29350 (Human)
  • Protein/peptide name:
    PTP1C
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MKAGFWEEFESLQKQEVKNLHQRLEGQRPENKGKNRYKNILPFDHSRVILQGRDSNIPGSDYINA NYIKNQLLGPDENAKTYIASQGCLEATVNDFWQMAWQENSRVIVMTTREVEKGRNKCVPYWPEVG MQRAYGPYSVTNCGEHDTTEYKLRTLQVSPLDNGDLIREIWHYQYLSWPDHGVPSEPGGVLSFLD QINQRQESLPHAGPIIVHCSAGIGRTGTIIVIDMLMENISTKGLDCDIDIQKTIQMVRAQRSGMV QTEAQYKFIYVAIAQFIETTKKKLEVLQSQKGQESEYGNILEHHHHHH
  • Formulation:
    Supplied as a 0.2 um filtered solution of 25mM TrisHCl, 2mM , -ME, 1mM EDTA, 1mM DTT, 20% Glycerol, pH 7.5.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together