Order Entry
Sweden
ContactUsLinkComponent
Mouse GMFB (from E. coli)
Mouse GMFB (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse GMFB (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    Less than 1EU/ug of rMuGMF beta as determined by LAL method.
  • Protein/peptide name:
    GMFB
  • Purity:
    >97% pure by SDS-PAGE and HPLC analyses.
  • Molecular weight:
    16.4 kDa
  • Sequence:
    SESLVVCDVA EDLVEKLRKF RFRKETHNAA IIMKIDKDER LVVLDEELEG VSPDELKDEL PERQPRFIVY SYKYQHDDGR VSYPLCFIFS SPVGCKPEQQ MMYAGSKNKL VQTAELTKVF EIRNTEDLTE EWLREKLGFF H
  • Formulation:
    Sterile Filtered White lyophilized (freeze-dried) powder. We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together