Human CX3CL1 (from E. coli)
: Biorbyt
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:Fully biologically active when compared to standard. Determined by its ability to chemoattract human T-Lymphocytes using a concentration range of 5.0-10.0 ng/ml. Less than 1EU/ug of rHuFractalkine/CX3CL1 as determined by LAL method.
- Protein synonyms:Fractalkine
- Protein/peptide name:CX3CL1
- Purity:>97% pure by SDS-PAGE and HPLC analyses.
- Molecular weight:8.5 kDa
- Sequence:QHHGVTKCNI TCSKMTSKIP VALLIHYQQN QASCGKRAII LETRQHRLFC ADPKEQWVKD AMQHLDRQAA ALTRNG
- Formulation:Lyophilized from a 0.2 um filtered concentrated (1.0mg/ml) solution in 20mM PB, pH 7.4, 50mM NaCl. We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom.
- Tested applications:SDS-PAGE , HPLC
Frequently Bought Together