Order Entry
Sweden
ContactUsLinkComponent
Cystatin recombinant Cystatin C (from E. coli)
Cystatin recombinant Cystatin C (from E. coli)
Catalog # BIRBORB81875-1
Supplier:  Biorbyt
CAS Number:  
Cystatin recombinant Cystatin C (from E. coli)
Catalog # BIRBORB81875-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Cystatin
  • Protein synonyms:
    AD8
  • Protein/peptide name:
    Cystatin C
  • Formulation:
    Lyophilised
  • Tested applications:
    ELISA
  • Purification:
    Affinity purified

Specifications

About this item