Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Apo E3 (from
E. coli
)
Recombinant Apo E3 (from E. coli)
Recombinant Apo E3 (from E. coli)
Catalog # BIRBORB81885-0.5
Supplier:
Biorbyt
CAS Number:
Expand image
Recombinant Apo E3 (from E. coli)
Catalog # BIRBORB81885-0.5
Supplier:
Biorbyt
CAS Number:
Specifications
Pk:
0,5 mg
Protein/peptide type:
Recombinant
Source:
E. coli
Protein/peptide name:
Apo E3
Purity:
Purified.
Formulation:
Lyophilised.
Specifications
About this item
Product Details
Documentation
Product Details
Documentation