Order Entry
Sweden
ContactUsLinkComponent
Mouse SDF1 alpha (from E. coli)
Mouse SDF1 alpha (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse SDF1 alpha (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The specific activity as determined by its ability to chemoattract human peripheral blood monocytes at 50-100 ng/ml.
  • Protein synonyms:
    C-X-C motif chemokine 12
  • Protein/peptide name:
    SDF1 alpha
  • Purity:
    > 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Stromal Cell-Derived Factor-1a in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Lys-Pro-Val-Ser-Leu.
  • Formulation:
    The CXCL12 protein was lyophilized from concentrated (1 mg/ml) sterile solution containing 10mM acetic acid (AcOH).
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together