Human A1M (from Urine (patients with chronic renal tubular proteinuria))
: Biorbyt
- Source:Urine (patients with chronic renal tubular proteinuria)
- Species:Human
- Storage conditions:Human A1M although stable at room temperature for weeks, should be stored between 2-8 °C.
- Protein synonyms:alpha-1 Microglobulin
- Protein/peptide name:A1M
- Purity:Greater than 96.0%.
- Solubility:Use phosphate buffer, pH>7.0 containing 0.15M NaCl, is recommended.
- Formulation:Lyophilized from 0.02M NH4C O3.
- Tested applications:SDS-PAGE
Frequently Bought Together