Human recombinant CD4 (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store vial at −20 °C to -80oC. When stored at the recommended temperature, this protein is stable for One year. Please avoid freeze-thaw cycles.
- Protein synonyms:CD 4
- Protein/peptide name:CD4
- Formulation:Recombinant Human protein at 100ug/ml in 50mM Tris-Acetate, pH-7.5, 1mM EDTA and 20% Glycerol.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together