Order Entry
Sweden
ContactUsLinkComponent
Human recombinant GDNF
Human recombinant GDNF
Supplier:  Biorbyt
Certificates
Human recombinant GDNF
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C.
  • Biological activity:
    The ED50 was determined by the proliferation of rat C6 cells is < 0.1 ng/ml, corresponding to a specific activity of > 1 x 107 units/mg.
  • Protein synonyms:
    Glial-Derived Neurotrophic Factor
  • UniProtKB:
    P39905
  • Protein/peptide name:
    GDNF
  • Purity:
    >98%
  • Solubility:
    It is recommended to reconstitute the lyophilized Glial Derived Neurotrophic Factor in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI
  • Formulation:
    GDNF was lyophilized after dialysis against 10mM sodium citrate and 150mM NaCl.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together