Order Entry
Sweden
ContactUsLinkComponent
Wortmannin, Millipore®
Wortmannin, Millipore®
Catalog # 681675-1
CAS Number:  
Wortmannin, Millipore®
Catalog # 681675-1
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :428.44 g/mol
CAS Number:19545-26-7
Formula:C₂₃H₂₄O₈
MDL Number:MFCD00133927