Jugs, HDPE, heavy duty, Nalgene®
Catalog # NALG642013-0710
Leverantör: Thermo Fisher Scientific
Specifikationer
- Capacity:20 l
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:NAR Certificate of Retirement
- Pk:4
Specifikationer
Om denna produkt
Made of HDPE.
- Good chemical resistance
- Good temperature resistance
- Sturdy