About this item
This MAb reacts with the beta-chain of HLA-DRB1 antigen, a member of MHC class II molecules. It does not cross react with HLA-DP and HLA-DQ. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36 kDa alpha (heavy) chain and a 28 kDa beta (light) chain. It is expressed on B-cells, activated T-cells, monocytes/macrophages, dendritic cells and other non-professional APCs. In conjunction with the CD3/TCR complex and CD4 molecules, HLA-DR is critical for efficient peptide presentation to CD4 T cells. It is an excellent histiocytic marker in paraffin sections producing intense cytoplasmic staining. True histiocytic neoplasms are similarly positive. HLA-DR antigens also occur on a variety of epithelial cells and their corresponding neoplastic counterparts. Loss of HLA-DR expression is related to tumor microenvironment and predicts adverse outcome in diffuse large B-cell lymphoma.
- Purified
Type: Primary
Antigen: HLA-DRB1
Clonality: Monoclonal
Clone: LN-3 HLA-DRB/1067
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Monkey
Specifications
- Catalog No:
- BNUB1208-500
- BNUB1208-100
- Antigensymbol:
- HLA-DRB1
- HLA-DRB1
- Antikroppsnamn:
- HLA-DRB
- HLA-DRB
- Antikroppstyp:
- Primär
- Primär
- Klonalitet:
- Monoclonal
- Monoclonal
- Konjugation:
- Unconjugated
- Unconjugated
- Klon:
- Clone LN-3 HLA-DRB/1067
- Clone LN-3 HLA-DRB/1067
- Reaktivitet:
- Monkey, Human
- Monkey, Human
- Värd:
- Mouse
- Mouse
- Gen-ID:
- 436568
- 436568
- Isotyp:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunKemi:
- Yes
- Yes
- Immunutfällning:
- Yes
- ImmunoFluorescens:
- Yes
- Yes
- Koncentration:
- 0,2 mg/ml
- 0,2 mg/ml
- Storage buffer:
- PBS/0,05% BSA/0,05% azide
- PBS/0,05% BSA/0,05% azide