Anti-SIGLEC12 Rabbit Polyclonal Antibody
Catalog # ABGEAP1630A
Supplier: Abgent
Specifications
- Antigen name:Sialic acid-binding Ig-like lectin 12
- Antigen symbol:SIGLEC12
- Clonality:Polyclonal
- Host:Rabbit
- Epitope:C-Terminal
- Antigen synonyms:mSiglec-E|Siglec9|SIGLECL1|Siglec5|Siglec12|Cd170|Siglecl1
- Size:100 µg
- Pk:400 µl
Specifications
About this item
Type:
Antigen: SIGLEC12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: