Anti-P62 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-42821R
Supplier: Novus Biologicals
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Nucleoporin p62
- Antigensymbol:P62
- Klonalitet:Polyclonal
- Konjugation:DyLight® 550
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Reaktivitet:Human
- Western blot:Yes
- Gen-ID:8878
- Antigen synonymer:ORCA|OSIL|SQSTM1|EBI3-associated protein of 60 kDa|EBIAP|p60|Ubiquitin-binding protein p62
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Storlek:100 μL
- Förp.:0,1 mL
Specifications
About this item
Rabbit Polyclonal p62/SQSTM1 Antibody [DyLight 550]. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human, Mouse, Rat.
Type: Primary
Antigen: P62
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human