Order Entry
Sweden
ContactUsLinkComponent
 
Anti-HLA-B Mouse Monoclonal Antibody (Purified) [clone: JOAN-1]
 :  Biotium
 
Anti-HLA-B Mouse Monoclonal Antibody (Purified) [clone: JOAN-1]
 :  Biotium

 

  • Catalog No:
  • BNUB1005-100
  • BNUB1005-500
  • Antigen symbol:
  • HLA-B
  • HLA-B
  • Antigen synonyms:
  • HLA-B|HLAB|Leukocyte antigen class I-B|MHC Class I HLA heavy chain
  • HLA-B|HLAB|Leukocyte antigen class I-B|MHC Class I HLA heavy chain
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone JOAN-1
  • Clone JOAN-1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 534332
  • 534332
  • Isotype:
  • IgG3 kappa
  • IgG3 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide