Anti-XRCC3 Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: XRCC3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP083-05
- DIABRP083
- Antigen symbol:
- XRCC3
- XRCC3
- Antigen name:
- X-ray Repair Complementing Defective Repair in Chinese Hamster cells 3
- X-ray Repair Complementing Defective Repair in Chinese Hamster cells 3
- Antigen synonyms:
- X ray repair complementing defective repair in Chinese hamster cells 3|CMM6|X ray repair cross complementing protein 3|DNA repair protein XRCC3
- X ray repair complementing defective repair in Chinese hamster cells 3|CMM6|X ray repair cross complementing protein 3|DNA repair protein XRCC3
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit