Anti-TDT Mouse Monoclonal Antibody [clone: SEN28]
Catalog # DIABPDM096
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:Terminal-deoxynucleotidyl transferase
- Antigen symbol:TDT
- Clonality:Monoclonal
- Clone:SEN28
- Host:Mouse
- Isotype:IgG2a kappa
- Antigen synonyms:Terminal-deoxynucleotidyl Transferase
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: TDT
Clonality: Monoclonal
Clone: SEN28
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: