Anti-T Cell Mouse Monoclonal Antibody [clone: UCHL1]
Catalog # DIABPDM015
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen symbol:T Cell
- Clonality:Monoclonal
- Clone:UCHL1
- Host:Mouse
- Isotype:IgG2a kappa
- Antigen synonyms:or transformation-related protein 53 (TRP53)|tumor suppressor p53|antigen NY-CO-13|P53|BCC7|TRP53|phosphoprotein p53|LFS1|tumor protein p53
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: T Cell
Clonality: Monoclonal
Clone: UCHL1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: