Anti-TGFB3 Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: TGFB3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP148-05
- DIABRP148
- Antigen symbol:
- TGFB3
- TGFbeta3
- Antigen name:
- Transforming Growth Factor Beta 3
- Antigen synonyms:
- ARVD, ARVD1, RNHF, TGF-beta3
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit