Anti-Synuclein (Pan) Rabbit Polyclonal Antibody
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Type:
Antigen: Synuclein (Pan)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifikationer
- Catalog No:
- DIABRP131-05
- DIABRP131
- Antigen name:
- pan Synuclein
- pan Synuclein
- Antigen synonyms:
- SNCB|beta-synuclein|Beta-synuclein.
- SNCB|beta-synuclein|Beta-synuclein.
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit