Anti-SYP Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: SYP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP036
- DIABRP036-05
- Antigen symbol:
- SYP
- SYP
- Antigen name:
- Synaptophysin
- Synaptophysin
- Antigen synonyms:
- MRXSYP|major synaptic vesicle protein P38|MRX96
- MRXSYP|major synaptic vesicle protein P38|MRX96
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit